Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium Chloride, Crystal, U.S.P., Multi-Compendial , J.T.Baker®

Supplier: Avantor Performance Materials

3045-09 3045-01 3045-07 3045-05
Crystals. Pyrogen tested. GMP manufactured.


Formula: KCl
MW: 74,55 g/mol
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704
Specification Test Results
USP - Assay (KCl) (dried basis) 99.0 - 100.5 %
USP - Acidity or Alkalinity Passes Test
USP - Bromide (Br) <= 0.1 %
USP - Calcium and Magnesium Passes Test
USP - Endotoxin Concentration (EU/g) <= 2.5
USP - Heavy Metals (as Pb) <= 0.001 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Iodides (I) <= 0.005 %
USP - Loss on Drying <= 1.00 %
USP - Sodium (Na) Passes Test
EP/BP - Assay (KCl) (dried basis) 99.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Bromide (Br) <= 0.1 %
EP/BP - Iodide (I) Passes Test
EP/BP - Sulfate (SO₄) <= 300 ppm
EP/BP - Barium (Ba) Passes Test
EP/BP - Heavy Metals (as Pb) <= 10 ppm
EP/BP - Iron (Fe) <= 20 ppm
EP/BP - Magnesium and Alkaline Earth Metals <= 200 ppm
EP/BP - Loss on Drying at 105°C <= 1.0 %
JP - Assay (KCl) (dried basis) 99 - 101 %
JP - Identification A Passes Test
JP - Identification B Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Acidity or Alkalinity Passes Test
JP - Bromide (Br) Passes Test
JP - Iodide (I) Passes Test
JP - Heavy Metals (as Pb) <= 5 ppm
JP - Calcium and Magnesium Passes Test
JP - Sodium (Na) Passes Test
JP - Arsenic (As) <= 2 ppm
JP - Loss on Drying <= 0.5 %


Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
500 g white poly bottle,blue T/E cap 3045-01 JT3045-1 Case of 4 (500 g) Retrieving Restricted
Each(500g) Retrieving Restricted
2.5 kg white poly bottle,blue T/E cap 3045-05 JT3045-5 Case of 4 (2,500 ,g) Retrieving Restricted
Each(2,500,g) Retrieving Restricted
12 kg polyethylene pail: screw top 3045-07 JT3045-7 Each (12kg) Retrieving Restricted
12 kg polyethylene pail: screw top 3045-07 MK304507 Each (12kg) Retrieving Restricted
100 lbs. open head polyethylene drum 3045-09 JT3045-9 Each (100lb) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty