Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 30% Ph. Eur., BP, USP

Supplier: MilliporeSigma

1.08597.2500 1.08597.1000 1.08597.9950 1.08597.9025
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Certifications: Corresponds to Ph. Eur., BP, USP.


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: 107 °C (1013 hPa)
Melting Pt: -26 °C
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00011333
CAS Number: 7722-84-1
UN: 2014
ADR: 5.1,II
Specification Test Results
pH value 2 - 4 (H₂O, 20 °C)
Vapor pressure 18 hPa (20 °C)
Storage Store at 2 to 25 °C
Assay (manganometric) 29.0 - 31.0%
Identity Passes Test
Acidity (according to Ph. Eur.) Passes Test
Acidity (according to USP) Passes Test
Chloride (Cl) ≤0.005%
Phosphate (PO₄) ≤0.05%
Heavy metals (as Pb) ≤0.0005%
As (Arsenic) ≤0.0003%
Cu (Copper) ≤0.002%
Pb (Lead) ≤0.001%
Pd (Palladium)* ≤0.0001%
Pt (Platinum)* ≤0.0001%
Zn (Zinc) ≤0.002%
Residual solvents (ICH Q3C) Excluded by production process
Organic stabilizers ≤0.045%
Non volatile matter (105 °C) ≤0.018%
Preservatives 0.015% Na₂H₂P₂O₇; 0.01% H₃PO₄; 0.006% NH₄NO₃

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
1 L Poly Bottle 1.08597.1000 EM1.08597.1000 Each (1l) Retrieving Restricted
2.5 L Poly Bottle 1.08597.2500 EM1.08597.2500 Each (2l) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 L Plastic Container 1.08597.9025 EM1.08597.9025 Each (25l) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

950 L Plastic IBC 1.08597.9950 EM1.08597.9950 Each (950l) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty