L(-)-Xylose ≥97.0% (by titrimetric analysis)

Supplier: TCI America

X0021-5G X0021-25G
TCX0021-005GEA 68.01 USD
TCX0021-005G TCX0021-25G
L(-)-Xylose ≥97.0% (by titrimetric analysis)
L(-)-Xylose

CAS Number: 609-06-3
MDL Number: MFCD00151096
Molecular Formula: C5H10O5
Molecular Weight: 150.13
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 150
Specific rotation [a]20/D: -20 deg (C=10, H2O)

Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00151096
CAS Number: 609-06-3
EINECS: 210-174-1
https://us.vwr.com/stibo/web/std.lang.all/73/82/28997382.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR