5-Undecanol ≥98.0%
Supplier: TCI America
Synonyms:
5-Undecyl alcohol
, 5-Hydroxy undecane
U0039-5ML
TCU0039-005MLEA
113.63
USD
TCU0039-005ML
5-Undecanol ≥98.0%
5-Undecanol
CAS Number: 37493-70-2
MDL Number: MFCD00046719
Molecular Formula: C11H24O
Molecular Weight: 172.31
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 140
Specific Gravity (20/20): 0.83
Formula:
C₁₁H₂₄O MW: 172.31 g/mol Boiling Pt: 140 °C Density: 0.83 (20 °C) |
MDL Number:
MFCD00046719 CAS Number: 37493-70-2 EINECS: 253-529-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...