3-Undecyne ≥96.0%

Supplier: TCI America

U0034-5ML
TCU0034-005MLEA 84 USD
TCU0034-005ML
3-Undecyne ≥96.0%
3-Undecyne

CAS Number: 60212-30-8
MDL Number: MFCD00041663
Molecular Formula: C11H20
Molecular Weight: 152.28
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Flash Point (°C): 65
Specific Gravity (20/20): 0.78

Formula: C₁₁H₂₀
Boiling Pt: 80 °C
Density: 0.78 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00041663
CAS Number: 60212-30-8

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR