1-Undecyne ≥96.0%

Supplier: TCI America

U0033-5ML
TCU0033-005MLEA 104 USD
TCU0033-005ML
1-Undecyne ≥96.0%
1-Undecyne

CAS Number: 2243-98-3
MDL Number: MFCD00015063
Molecular Formula: C11H20
Molecular Weight: 152.28
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 195
Flash Point (°C): 65
Specific Gravity (20/20): 0.77


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Rutylidene

Formula: C₁₁H₂₀
MW: 152.28 g/mol
Boiling Pt: 195 °C
Melting Pt: –25 °C
Density: 0.77 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00015063
CAS Number: 2243-98-3
EINECS: 218-825-1

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR