Triphenylethylene ≥98.0%
Supplier: TCI America
Synonyms:
1,1,2-Triphenylethylene
, 1,1,2-Triphenylethene
T1834-25G
TCT1834-025GEA
94.36
USD
TCT1834-025G
Triphenylethylene ≥98.0%
Triphenylethylene
CAS Number: 58-72-0
MDL Number: MFCD00004765
Molecular Formula: C20H16
Molecular Weight: 256.35
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 69
Formula:
(C₆H₅)₂CCHC₆H₅ MW: 256.35 g/mol Boiling Pt: 358 °C (1013 hPa) Melting Pt: 69 °C Density: 1.0373 g/cm³ (78.4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004765 CAS Number: 58-72-0 EINECS: 200-395-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...