Tetrabromohydroquinone ≥97.0%

Supplier: TCI America

T1790-25G T1790-5G
TCT1790-005GEA 60.88 USD
TCT1790-005G TCT1790-025G
Tetrabromohydroquinone ≥97.0%
Tetrabromohydroquinone

CAS Number: 2641-89-6
MDL Number: MFCD00013970
Molecular Formula: C6H2Br4O2
Molecular Weight: 425.70
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Color: Very Pale Yellow


Other Product Names / Synonyms: 1,2,4,5-Tetrabromo-3,6-dihydroxybenzene, Tetrabromoquinol

Formula: C₆H₂Br₄O₂
MDL Number: MFCD00013970
CAS Number: 2641-89-6
EINECS: 220-142-9
https://us.vwr.com/stibo/web/std.lang.all/57/67/28995767.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR