3-Hydroxy-2,4,6-tribromobenzoic acid ≥95.0% (by titrimetric analysis) for biochemical research
Supplier: TCI America
Synonyms:
2,4,6-Tribromo-3-hydroxybenzoic acid
T1764-5G
TCT1764-005GEA
45
USD
TCT1764-005G
3-Hydroxy-2,4,6-tribromobenzoic acid ≥95.0% (by titrimetric analysis) for biochemical research
3-Hydroxy-2,4,6-tribromobenzoic acid
[for Biochemical Research]
CAS Number: 14348-40-4
MDL Number: MFCD00055557
Molecular Formula: C7H3Br3O3
Molecular Weight: 374.81
Purity/Analysis Method:
Form: Crystal
Color: White
Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: TBHBA
Formula:
C₇H₃Br₃O₃ Boiling Pt: 396 °C (1013 hPa) Melting Pt: 146 °C |
MDL Number:
MFCD00055557 CAS Number: 14348-40-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...