1,5,5-Trimethylhydantoin ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1739-25G
TCT1739-025GEA 151 USD
TCT1739-025G
1,5,5-Trimethylhydantoin ≥98.0% (by titrimetric analysis)
1,5,5-Trimethylhydantoin

CAS Number: 6851-81-6
MDL Number: MFCD00040439
Molecular Formula: C6H10N2O2
Molecular Weight: 142.16
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 164


Other Product Names / Synonyms: 1,5,5-Trimethylimidazolidine-2,4-dione

Formula: C₆H₁₀N₂O₂
Melting Pt: 164 °C
MDL Number: MFCD00040439
CAS Number: 6851-81-6
EINECS: 229-945-9
https://us.vwr.com/stibo/web/std.lang.all/57/20/28995720.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR