4-Tritylphenol ≥96.0%
Supplier: TCI America
T1654-5G
TCT1654-005GEA
137
USD
TCT1654-005G
4-Tritylphenol ≥96.0%
4-Tritylphenol
CAS Number: 978-86-9
MDL Number: MFCD00002364
Molecular Formula: C25H20O
Molecular Weight: 336.43
Purity/Analysis Method:
Form: Crystal
Color: White
Other Product Names / Synonyms: 4-Hydroxytetraphenylmethane, 4-Tritylphenol
Formula:
C₂₅H₂₀O |
MDL Number:
MFCD00002364 CAS Number: 978-86-9 EINECS: 213-557-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...