TCIAT1644

T1644-5G
TCT1644-005GEA 172.56 USD
TCT1644-005G
TCIAT1644
CAS Number: 5044-52-0
MDL Number: MFCD00011807
Molecular Formula: C20H18BrP
Molecular Weight: 369.24
Purity/Analysis Method: 96.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 187

Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Vinylation
Other Product Names / Synonyms: Schweizer's Reagent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR