Triethyloxonium tetrafluoroborate 15% ∼1 mol/L in dichloromethane
Supplier: TCI America
T1606-100ML
TCT1606-100MLEA
149.21
USD
TCT1606-100ML
Triethyloxonium tetrafluoroborate 15% ∼1 mol/L in dichloromethane
Triethyloxonium tetrafluoroborate
[Ethylating Reagent]
CAS Number: 368-39-8
MDL Number: MFCD00044423
Molecular Formula: C6H15BF4O
Molecular Weight: 189.99
Form: Clear Liquid
Color: Colorless
Storage Temperature:
Formula:
C₆H₁₅BF₄O |
MDL Number:
MFCD00044423 CAS Number: 368-39-8 EINECS: 206-705-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...