Lithium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 33454-82-9
MDL Number: MFCD00013227
Molecular Formula: CHF3O3S
Molecular Weight: 156.00
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 423
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Lithium Triflate, Trifluoromethanesulfonic Acid Lithium Salt
Formula:
CF₃LiO₃S Melting Pt: 423 °C |
MDL Number:
MFCD00013227 CAS Number: 33454-82-9 EINECS: 251-528-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...