α,α,α-Trifluoro-o-toluic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(Trifluoromethyl)benzoic acid

T1302-10G
TCT1302-010GEA 57.86 USD
TCT1302-010G
α,α,α-Trifluoro-o-toluic acid ≥98.0% (by HPLC, titration analysis)
α,α,α-Trifluoro-o-toluic acid

CAS Number: 433-97-6
MDL Number: MFCD00002476
Molecular Formula: C8H5F3O2
Molecular Weight: 190.12
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Boiling point (°C): 247
Melting point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha-Trifluoro-o-toluic Acid

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 247 °C
Melting Pt: 110 °C
MDL Number: MFCD00002476
CAS Number: 433-97-6
EINECS: 207-093-9
https://us.vwr.com/stibo/web/std.lang.all/53/37/28995337.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR