Tin (II) trifluoromethanesulfonate ≥90.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Tin trifluoromethanesulfonate
, Tin(II) trifluoromethanesulfonate
, Tin triflate
, Tin(II) trifluoromethanesulphonate
, Tin trifluoromethanesulphonate
CAS Number: 62086-04-8
MDL Number: MFCD00191251
Molecular Formula: CHF3O3S
Molecular Weight: 416.84
Purity/Analysis Method:
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts
Other Product Names / Synonyms: Stannous Trifluoromethanesulfonate, Tin(II) Triflate, Trifluoromethanesulfonic Acid Tin(II) Salt
Formula:
C₂F₆O₆S₂Sn MW: 416.85 g/mol |
MDL Number:
MFCD00191251 CAS Number: 62086-04-8 UN: 3288 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...