Tetrapropoxysilane ≥98.0%

Supplier: TCI America
Warning

Synonyms: Tetrapropyl orthosilicateTetra-n-propoxysilane

T1174-25ML T1174-500ML
TCT1174-500MLEA 178.59 USD
TCT1174-500ML TCT1174-25ML
Tetrapropoxysilane ≥98.0%
Tetrapropoxysilane

CAS Number: 682-01-9
MDL Number: MFCD00009364
Molecular Formula: C12H28O4Si
Molecular Weight: 264.44
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 225
Flash Point (°C): 104
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Tetrapropoxysilane

Formula: C₁₂H₂₈O₄Si
MW: 264.44 g/mol
Boiling Pt: 225 °C
Density: 0.91 (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00009364
CAS Number: 682-01-9
EINECS: 211-659-0
https://us.vwr.com/stibo/web/std.lang.all/52/26/28995226.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR