1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) ≥95.0%
Supplier: TCI America
Synonyms:
Beta-tetralone
T1167-5G
T1167-25G
TCT1167-005GEA
353
USD
TCT1167-005G
TCT1167-25G
1,2,3,4-Tetrahydronaphthalen-2-one (2-tetralone-β-tetralone) ≥95.0%
1,2,3,4-Tetrahydronaphthalen-2-one (2-Tetralone, β-Tetralone)
CAS Number: 530-93-8
MDL Number: MFCD00001727
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 240
Flash Point (°C): 110
Freezing point (°C): 14
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: 3,4-Dihydro-2(1H)-naphthalenone
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Boiling Pt: 240 °C Density: 1.10 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001727 CAS Number: 530-93-8 EINECS: 208-498-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...