3,4,5-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (2E)-3-(3,4,5-Trimethoxyphenyl)acrylic acid

T1104-25G
TCT1104-025GEA 69 USD
TCT1104-025G
3,4,5-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
3,4,5-Trimethoxycinnamic acid

CAS Number: 90-50-6
MDL Number: MFCD00004388
Molecular Formula: C12H14O5
Molecular Weight: 238.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 129


Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(3,4,5-Trimethoxyphenyl)acrylic Acid

Formula: C₁₂H₁₄O₅
MW: 238.24 g/mol
Boiling Pt: 367 °C (1013 hPa)
Melting Pt: 129 °C
MDL Number: MFCD00004388
CAS Number: 90-50-6
EINECS: 201-999-8
https://us.vwr.com/stibo/web/std.lang.all/51/71/28995171.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR