Chlorthaldimethyl ≥96.0%
Supplier: TCI America
Synonyms:
Dimethyl tetrachloroterephthalate
T1064-25G
TCT1064-025GEA
49.74
USD
TCT1064-025G
Chlorthaldimethyl ≥96.0%
Chlorthal-dimethyl
CAS Number: 1861-32-1
MDL Number: MFCD00014902
Molecular Formula: C10H6Cl4O4
Molecular Weight: 331.95
Purity/Analysis Method:
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 156
Other Product Names / Synonyms: Tetrachloroterephthalic Acid Dimethyl Ester
Formula:
C₁₀H₆Cl₄O₄ MW: 331.97 Melting Pt: 156 °C |
MDL Number:
MFCD00014902 CAS Number: 1861-32-1 EINECS: 217-464-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...