2,3,5,6-Tetrafluoroaniline ≥97.0%
Supplier: TCI America
|
Warning
|
T1028-5G
T1028-25G
TCT1028-025GEA
149.21
USD
TCT1028-025G
TCT1028-005G
2,3,5,6-Tetrafluoroaniline ≥97.0%
2,3,5,6-Tetrafluoroaniline
CAS Number: 700-17-4
MDL Number: MFCD00007644
Molecular Formula: C6H3F4N
Molecular Weight: 165.09
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 158
Flash Point (°C): 62
Freezing point (°C): 31
Formula:
C₆H₃F₄N MW: 165.09 g/mol Boiling Pt: 158 °C Melting Pt: 31…32 °C Density: 1.744 g/cm³ (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00007644 CAS Number: 700-17-4 EINECS: 211-840-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...