Tryptamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(1H-indol-3-yl)ethan-1-amine

T0890-25G
TCT0890-025GEA 80.53 USD
TCT0890-025G
Tryptamine ≥98.0% (by HPLC, titration analysis)
Tryptamine

CAS Number: 61-54-1
MDL Number: MFCD00005661
Molecular Formula: C10H12N2
Molecular Weight: 160.22
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 117
Flash Point (°C): 185
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(2-Aminoethyl)indole

Formula: C₁₀H₁₂N₂
Melting Pt: 117 °C
Flash Pt: 185 °C
MDL Number: MFCD00005661
CAS Number: 61-54-1
EINECS: 200-510-5
https://us.vwr.com/stibo/web/std.lang.all/49/91/28994991.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR