m-Tolylacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(3-Methylphenyl)acetic acid
, 2-(m-Tolyl)acetic acid
T0880-25G
TCT0880-025GEA
44.66
USD
TCT0880-025G
m-Tolylacetic acid ≥98.0% (by titrimetric analysis)
m-Tolylacetic acid
CAS Number: 621-36-3
MDL Number: MFCD00004340
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 64
Other Product Names / Synonyms: 3-Methylphenylacetic Acid
Formula:
(C₆H₄(CH₃))CH₂COOH MW: 150.18 g/mol Boiling Pt: 120…123 °C (26 mmHg) Melting Pt: 64 °C |
MDL Number:
MFCD00004340 CAS Number: 621-36-3 EINECS: 210-683-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...