Bromotriphenylethylene ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-1,1,2-triphenylethylene

T0853-25G
TCT0853-025GEA 97 USD
TCT0853-025G
Bromotriphenylethylene ≥98.0% (by GC, titration analysis)
Bromotriphenylethylene

CAS Number: 1607-57-4
MDL Number: MFCD00000135
Molecular Formula: C20H15Br
Molecular Weight: 335.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 114

Formula: (C₆H₅)₂CCBrC₆H₅
MW: 335.24 g/mol
Boiling Pt: 413 °C (1013 hPa)
Melting Pt: 114 °C
MDL Number: MFCD00000135
CAS Number: 1607-57-4
EINECS: 216-531-8
https://us.vwr.com/stibo/web/std.lang.all/49/62/28994962.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR