Tetramethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Danger

Synonyms: Tetramethylammoniumtetrafluoroborate

T0842-500G T0842-25G
TCT0842-500GEA 565.1 USD
TCT0842-500G TCT0842-25G
Tetramethylammonium tetrafluoroborate ≥98.0% (by total nitrogen basis)
Tetramethylammonium tetrafluoroborate

CAS Number: 661-36-9
MDL Number: MFCD00011745
Molecular Formula: C4H12BF4N
Molecular Weight: 160.95
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 330

Formula: C₄H₁₂BF₄N
MW: 160.95 g/mol
Melting Pt: 330 °C
MDL Number: MFCD00011745
CAS Number: 661-36-9
EINECS: 211-547-1
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/52/28994952.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR