Phenyl trifluoroacetate ≥96.0%

Supplier: TCI America

Synonyms: Trifluoroacetic phenyl ester

T0741-25G
TCT0741-025GEA 55.83 USD
TCT0741-025G
Phenyl trifluoroacetate ≥96.0%
Phenyl trifluoroacetate

CAS Number: 500-73-2
MDL Number: MFCD00039232
Molecular Formula: C8H5F3O2
Molecular Weight: 190.12
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 148
Flash Point (°C): 52
Specific Gravity (20/20): 1.29


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Trifluoroacetic Acid Phenyl Ester

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 148 °C
Melting Pt: –8.5 °C
Density: 1.29 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00039232
CAS Number: 500-73-2
EINECS: 207-911-4
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/48/80/28994880.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR