1,2,3,4-Tetrahydronaphthalene ≥98.0%

Supplier: TCI America
Warning

T0713-250ML
TCT0713-250MLEA 95.41 USD
TCT0713-250ML
1,2,3,4-Tetrahydronaphthalene ≥98.0%
1,2,3,4-Tetrahydronaphthalene

[for Spectrophotometry]
CAS Number: 119-64-2
MDL Number: MFCD00001733
Molecular Formula: C10H12
Molecular Weight: 132.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 207
Flash Point (°C): 75
Specific Gravity (20/20): 0.97

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 207 °C
Melting Pt: –35.8 °C
Density: 0.97 (20 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00001733
CAS Number: 119-64-2
EINECS: 204-340-2
UN: 3082
ADR: 9,III
Merck Index: 12,09360
https://us.vwr.com/stibo/web/std.lang.all/48/65/28994865.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR