Methyl trifluoroacetate ≥98.0%

Supplier: TCI America

Synonyms: Trifluoroacetic methyl ester

T0680-25G
TCT0680-25GEA 19.27 USD
TCT0680-25G
Methyl trifluoroacetate ≥98.0%
Methyl trifluoroacetate

CAS Number: 431-47-0
MDL Number: MFCD00000417
Molecular Formula: C3H3F3O2
Molecular Weight: 128.05
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 43
Melting point (°C): -78
Flash Point (°C): -20
Specific Gravity (20/20): 1.29


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Trifluoroacetic Acid Methyl Ester

Formula: C₃H₃F₃O₂
MW: 128.05 g/mol
Boiling Pt: 43 °C
Melting Pt: –78 °C
Density: 1.29 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00000417
CAS Number: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/48/46/28994846.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR