TCIAT0610

T0610-25ML
TCT0610-25MLEA 57.35 USD
TCT0610-25ML
TCIAT0610
CAS Number: 116-17-6
MDL Number: MFCD00008874
Molecular Formula: C9H21O3P
Molecular Weight: 208.24
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 74
Flash Point (°C): 62
Specific Gravity (20/20): 0.91

Other Product Names / Synonyms: Phosphorous Acid Triisopropyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR