2,3,5-Trimethylphenol ≥98.0%

Supplier: TCI America

T0485-500G T0485-25G
TCT0485-025GEA 30.43 USD
TCT0485-025G TCT0485-500G
2,3,5-Trimethylphenol ≥98.0%
2,3,5-Trimethylphenol

CAS Number: 697-82-5
MDL Number: MFCD00002228
Molecular Formula: C9H12O
Molecular Weight: 136.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 233
Melting point (°C): 93


Other Product Names / Synonyms: Isopseudocumenol

Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 233 °C
Melting Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002228
CAS Number: 697-82-5
EINECS: 211-806-9
UN: 2430
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/47/37/28994737.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR