2,3,5-Trimethylhydroquinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2,3,5-Trimethylbenzene-1,4-diolTrimethylhydroquinone

T0477-25G T0477-500G
TCT0477-500GEA 406.57 USD
TCT0477-500G TCT0477-025G
2,3,5-Trimethylhydroquinone ≥98.0% (by GC, titration analysis)
2,3,5-Trimethylhydroquinone

CAS Number: 700-13-0
MDL Number: MFCD00002346
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 169
Flash Point (°C): 191

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 169 °C
Flash Pt: 191 °C
Storage Temperature: Ambient
MDL Number: MFCD00002346
CAS Number: 700-13-0
EINECS: 211-838-3
UN: 3077
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/47/31/28994731.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR