Mesitylene ≥97.0%

Supplier: TCI America
Danger

Synonyms: 1,3,5-Trimethylbenzene

T0470-500ML T0470-25ML
TCT0470-500MLEA 82.68 USD
TCT0470-500ML TCT0470-25ML
Mesitylene ≥97.0%
Mesitylene

CAS Number: 108-67-8
MDL Number: MFCD00008538
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 164
Flash Point (°C): 50
Specific Gravity (20/20): 0.87


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Mesitylene

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 164 °C
Melting Pt: –45 °C
Density: 0.87 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00008538
CAS Number: 108-67-8
EINECS: 203-604-4
UN: 2325
ADR: 3,III
Merck Index: 13,05934
https://us.vwr.com/stibo/web/std.lang.all/47/25/28994725.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR