Fentin chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Chlorotriphenylstannane
T0447-5G
TCT0447-005GEA
53.8
USD
TCT0447-005G
Fentin chloride ≥95.0% (by titrimetric analysis)
Fentin chloride
CAS Number: 639-58-7
MDL Number: MFCD00000519
Molecular Formula: C18H15ClSn
Molecular Weight: 385.48
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 105
Formula:
C₁₈H₁₅ClSn MW: 385.48 g/mol Boiling Pt: 397 °C (1013 hPa) Melting Pt: 105 °C Density: 1.49 g/cm³ (20 °C) Flash Pt: >150 °C |
MDL Number:
MFCD00000519 CAS Number: 639-58-7 EINECS: 211-358-4 UN: 3146 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...