2-Tridecanone ≥95.0%

Supplier: TCI America

T0415-25G
TCT0415-025GEA 54.81 USD
TCT0415-025G
2-Tridecanone ≥95.0%
2-Tridecanone

CAS Number: 593-08-8
MDL Number: MFCD00008968
Molecular Formula: C13H26O
Molecular Weight: 198.35
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 263
Flash Point (°C): 107
Freezing point (°C): 27


Other Product Names / Synonyms: Methyl Undecyl Ketone

Formula: C₁₃H₂₆O
MW: 198.35 g/mol
Boiling Pt: 263 °C
Melting Pt: 25…27 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008968
CAS Number: 593-08-8
EINECS: 209-784-0
https://us.vwr.com/stibo/web/std.lang.all/46/85/28994685.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR