α,3,4-Trichlorotoluene ≥98.0%

Supplier: TCI America
Danger

Synonyms: 1,2-Dichloro-4-(chloromethyl)benzene1-(Chloromethyl)-3,4-dichlorobenzene3,4-Dichlorobenzyl chloride

T0401-500G T0401-25G
TCT0401-25GEA 28.42 USD
TCT0401-25G TCT0401-500G
α,3,4-Trichlorotoluene ≥98.0%
α,3,4-Trichlorotoluene

CAS Number: 102-47-6
MDL Number: MFCD00000906
Molecular Formula: C7H5Cl3
Molecular Weight: 195.47
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 241
Flash Point (°C): 155
Specific Gravity (20/20): 1.42


Other Product Names / Synonyms: alpha,3,4-Trichlorotoluene

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 241 °C
Melting Pt: 37.5 °C
Density: 1.42 (20 °C)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00000906
CAS Number: 102-47-6
EINECS: 203-033-0
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/46/74/28994674.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR