Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,2,4-Trichlorotoluene ≥98.0%

Supplier: TCI America

Danger
Synonyms: 1,3-Dichloro-4-(chloromethyl)benzene1-(Chloromethyl)-2,4-dichlorobenzene2,4-Dichlorobenzyl chloride

T0400-500G T0400-25G
TCT0400-500GEA 134.21 USD InStock
TCT0400-500G TCT0400-25G
α,2,4-Trichlorotoluene ≥98.0%
α,2,4-Trichlorotoluene
CAS Number: 94-99-5
MDL Number: MFCD00000895
Molecular Formula: C7H5Cl3
Molecular Weight: 195.47
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 248
Flash Point (°C): 110
Specific Gravity (20/20): 1.41

Other Product Names / Synonyms: alpha,2,4-Trichlorotoluene


Formula: C₇H₅Cl₃
MW: 195.48 g/mol
Boiling Pt: 248 °C
Melting Pt: -2,7 °C
Density: 1.406 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000895
CAS Number: 94-99-5
EINECS: 202-381-0
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty