Homophthalic acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-(Carboxymethyl)benzoic acid
T0262-250G
T0262-25G
TCT0262-250GEA
219.3
USD
TCT0262-250G
TCT0262-025G
Homophthalic acid ≥99.0% (by titrimetric analysis)
Homophthalic acid
CAS Number: 89-51-0
MDL Number: MFCD00004326
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 181
Other Product Names / Synonyms: 2-Carboxyphenylacetic Acid
Formula:
C₉H₈O₄ MW: 180.16 g/mol Melting Pt: 181 °C |
MDL Number:
MFCD00004326 CAS Number: 89-51-0 EINECS: 201-913-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...