Methyl tiglate ≥98.0%

Supplier: TCI America

T0248-100ML T0248-25ML
TCT0248-100MLEA 218 USD
TCT0248-100ML TCT0248-025ML
Methyl tiglate ≥98.0%
Methyl tiglate

CAS Number: 6622-76-0
MDL Number: MFCD00016654
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 139
Flash Point (°C): 39
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Tiglic Acid Methyl Ester

Formula: C₆H₁₀O₂
Boiling Pt: 139 °C
Density: 0.95 (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00016654
CAS Number: 6622-76-0
EINECS: 229-575-8
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/45/50/28994550.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR