Ethyl tiglate ≥98.0%

Supplier: TCI America

T0247-25G
TCT0247-025GEA 47.71 USD
TCT0247-025G
Ethyl tiglate ≥98.0%
Ethyl tiglate

CAS Number: 5837-78-5
MDL Number: MFCD00015183
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 156
Flash Point (°C): 50
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: Tiglic Acid Ethyl Ester

Formula: C₇H₁₂O₂
Boiling Pt: 156 °C
Density: 0.93 (20 °C)
Flash Pt: 50 °C
MDL Number: MFCD00015183
CAS Number: 5837-78-5
EINECS: 227-425-6
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/45/49/28994549.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR