Ethyl mercaptoacetate ≥97.0%

Supplier: TCI America

Synonyms: Ethyl thioglycolateEthyl 2-mercaptoacetate

T0211-500G T0211-25G
TCT0211-500GEA 228.38 USD
TCT0211-500G TCT0211-025G
Ethyl mercaptoacetate ≥97.0%
Ethyl mercaptoacetate

CAS Number: 623-51-8
MDL Number: MFCD00004874
Molecular Formula: C4H8O2S
Molecular Weight: 120.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 157
Melting point (°C): -80
Flash Point (°C): 48
Specific Gravity (20/20): 1.10


Other Product Names / Synonyms: Ethyl Mercaptoacetate, Mercaptoacetic Acid Ethyl Ester, Thioglycolic Acid Ethyl Ester

Formula: C₄H₈O₂S
MW: 120.17 g/mol
Boiling Pt: 157 °C
Melting Pt: –80 °C
Density: 1.10 (20 °C)
Flash Pt: 48 °C
MDL Number: MFCD00004874
CAS Number: 623-51-8
EINECS: 210-800-3
https://us.vwr.com/stibo/web/std.lang.all/45/24/28994524.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR