2,2'-Thiodiacetic acid ≥98.0% (by GC, titration analysis)
|
Warning
|
Synonyms:
Thiodiacetic Acid
, Thiodiglycolic acid
T0203-500G
TCT0203-500GEA
144.2
USD
TCT0203-500G
2,2'-Thiodiacetic acid ≥98.0% (by GC, titration analysis)
2,2'-Thiodiacetic acid
CAS Number: 123-93-3
MDL Number: MFCD00004359
Molecular Formula: C4H6O4S
Molecular Weight: 150.15
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 129
Other Product Names / Synonyms: 2,2'-Thiodiacetic Acid
Formula:
C₄H₆O₄S MW: 150.16 g/mol Boiling Pt: 168 °C (14 mmHg) Melting Pt: 129 °C Density: 1.661 g/cm³ (20 °C) |
MDL Number:
MFCD00004359 CAS Number: 123-93-3 EINECS: 204-663-9 UN: 3261 ADR: 8,III Merck Index: 12,09467 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...