1,3-Diphenyl-2-thiourea

Supplier: TCI America

Synonyms: N,N'-Diphenylthiourea Thiocarbanilide

T0197-500G T0197-25G
TCT0197-500GEA 109.62 USD
TCT0197-500G TCT0197-25G
1,3-Diphenyl-2-thiourea
1,3-Diphenyl-2-thiourea

CAS Number: 102-08-9
MDL Number: MFCD00004921
Molecular Formula: C13H12N2S
Molecular Weight: 228.31
Form: Crystal
Color: White
Melting point (°C): 151


Other Product Names / Synonyms: Thiocarbanilide

Formula: C₁₃H₁₂N₂S
MW: 228.32 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.3205 g/cm³ (20 °C)
MDL Number: MFCD00004921
CAS Number: 102-08-9
EINECS: 203-004-2
UN: 2811
ADR: 6.1,II
Merck Index: 13,03366
https://us.vwr.com/stibo/web/std.lang.all/45/14/28994514.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR