Tetramethylammonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: TMACl

T0136-25G T0136-500G
TCT0136-500GEA 96.41 USD
TCT0136-500G TCT0136-25G
Tetramethylammonium chloride ≥98.0% (by titrimetric analysis)
Tetramethylammonium chloride

CAS Number: 75-57-0
MDL Number: MFCD00011628
Molecular Formula: C4H12ClN
Molecular Weight: 109.60
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White

Formula: (CH₃)₄NCl
MW: 109.60 g/mol
Melting Pt: > 300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/44/61/28994461.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR