Tetramethylammonium bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

T0135-500G T0135-25G
TCT0135-25GEA 16.24 USD
TCT0135-25G TCT0135-500G
Tetramethylammonium bromide ≥97.0% (by titrimetric analysis)
Tetramethylammonium bromide

CAS Number: 64-20-0
MDL Number: MFCD00011626
Molecular Formula: C4H12BrN
Molecular Weight: 154.05
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White

Formula: C₄H₁₂BrN
MW: 154.05 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/44/59/28994459.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR