1-Tetradecanethiol ≥97.0%

Supplier: TCI America

Synonyms: 1-Mercaptotetradecane n-Tetradecanethiol n-Tetradecyl mercaptan

T0082-25ML
TCT0082-025MLEA 87.29 USD
TCT0082-025ML
1-Tetradecanethiol ≥97.0%
1-Tetradecanethiol

CAS Number: 2079-95-0
MDL Number: MFCD00022099
Molecular Formula: C14H30S
Molecular Weight: 230.45
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 310
Flash Point (°C): 110
Freezing point (°C): 7
Specific Gravity (20/20): 0.85


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Myristyl Mercaptan, Tetradecyl Mercaptan

Formula: C₁₄H₃₀S
MW: 230.46 g/mol
Boiling Pt: 310 °C
Melting Pt: 7 °C
Density: 0.85 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00022099
CAS Number: 2079-95-0
EINECS: 218-209-2
UN: 3334
ADR: 9,
https://us.vwr.com/stibo/web/std.lang.all/44/17/28994417.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR