n-Tetracosane ≥99.0%
Supplier: TCI America
Synonyms:
N-tetracosane
T0075-25G
T0075-100G
T0075-500G
TCT0075-100GEA
179.66
USD
TCT0075-100G
TCT0075-500G
TCT0075-025G
n-Tetracosane ≥99.0%
n-Tetracosane
CAS Number: 646-31-1
MDL Number: MFCD00009352
Molecular Formula: C24H50
Molecular Weight: 338.66
Purity/Analysis Method:
Form: Crystal
Color: White
Boiling point (°C): 250
Melting point (°C): 51
Flash Point (°C): 194
Formula:
H₃C(CH₂)₂₂CH₃ MW: 338.66 g/mol Boiling Pt: 250 °C Melting Pt: 51 °C Density: 0.7787 g/cm³ (20 °C) Flash Pt: 194 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009352 CAS Number: 646-31-1 EINECS: 211-474-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...