α,α,α,4-Tetrachlorotoluene ≥98.0%

Synonyms: 4-Chlorobenzotrichloride1-Chloro-4-(trichloromethyl)benzene

T0072-500G T0072-25G
TCT0072-500GEA 65.49 USD
TCT0072-500G TCT0072-25G
α,α,α,4-Tetrachlorotoluene ≥98.0%
α,α,α,4-Tetrachlorotoluene

CAS Number: 5216-25-1
MDL Number: MFCD00000790
Molecular Formula: C7H4Cl4
Molecular Weight: 229.91
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 245
Melting point (°C): 6
Flash Point (°C): 140
Specific Gravity (20/20): 1.49


Other Product Names / Synonyms: alpha,alpha,alpha,4-Tetrachlorotoluene

Formula: C₇H₄Cl₄
MW: 229.92 g/mol
Boiling Pt: 245 °C
Melting Pt: 6 °C
Density: 1.49 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00000790
CAS Number: 5216-25-1
EINECS: 226-009-1
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/44/10/28994410.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR