1,1,2,2-Tetrachloroethane ≥97.0%
Supplier: TCI America
|
Danger
|
T0063-25G
T0063-500G
TCT0063-500GEA
65.34
USD
TCT0063-500G
TCT0063-25G
1,1,2,2-Tetrachloroethane ≥97.0%
1,1,2,2-Tetrachloroethane
CAS Number: 79-34-5
MDL Number: MFCD00000848
Molecular Formula: C2H2Cl4
Molecular Weight: 167.84
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 146
Melting point (°C): -44
Specific Gravity (20/20): 1.60
Other Product Names / Synonyms: Acetylene Tetrachloride
Formula:
Cl₂HCCHCl₂ MW: 167.85 g/mol Boiling Pt: 146 °C Melting Pt: –44 °C Density: 1.60 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000848 CAS Number: 79-34-5 EINECS: 201-197-8 UN: 1702 ADR: 6.1,II Merck Index: 12,09331 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...