1,1,2,2-Tetrabromoethane ≥98.0%
Supplier: TCI America
|
Danger
|
T0034-500G
T0034-25G
TCT0034-500GEA
95.75
USD
TCT0034-500G
TCT0034-25G
1,1,2,2-Tetrabromoethane ≥98.0%
1,1,2,2-Tetrabromoethane
CAS Number: 79-27-6
MDL Number: MFCD00000133
Molecular Formula: C2H2Br4
Molecular Weight: 345.65
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 243
Melting point (°C): 0
Specific Gravity (20/20): 2.97
Other Product Names / Synonyms: Acetylene Tetrabromide, TBE
Formula:
Br₂HCCHBr₂ MW: 345.65 g/mol Boiling Pt: 243 °C Melting Pt: –1 °C Density: 2.97 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000133 CAS Number: 79-27-6 EINECS: 201-191-5 UN: 2504 ADR: 6.1,III Merck Index: 12,09328 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...