o-Terphenyl ≥99.0%

Supplier: TCI America
Warning

Synonyms: 1,2-Diphenylbenzene 2-Phenyl-1,1'-biphenyl 1,2-Terphenyl 2-Phenylbiphenyl

T0019-100G T0019-500G T0019-25G
TCT0019-500GEA 624.04 USD
TCT0019-500G TCT0019-100G TCT0019-025G
o-Terphenyl ≥99.0%
o-Terphenyl

CAS Number: 84-15-1
MDL Number: MFCD00003055
Molecular Formula: C18H14
Molecular Weight: 230.31
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 332
Melting point (°C): 58
Flash Point (°C): 163
Lambda max.: 233 nm (Cyclohexane)


Product Categories: Materials Chemistry, Nanocarbon Unit Structures (Nanocarbon Materials)
Other Product Names / Synonyms: 1,2-Diphenylbenzene

Formula: C₁₈H₁₄
MW: 230.31 g/mol
Boiling Pt: 332 °C
Melting Pt: 58 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00003055
CAS Number: 84-15-1
EINECS: 201-517-6
https://us.vwr.com/stibo/web/std.lang.all/43/61/28994361.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR